Learn More
Medchemexpress LLC (R)-Thalidomide | 2614-06-4 | 99.8% | 258.23 | 1 MG
SDP
Catalog No. 5000363556
Encompass_Preferred
(R)-Thalidomide, also known as (R)-(+)-Thalidomide, is the R-enantiomer of Thalidomide, possessing psychomotor stabilizing properties. It is intended for research use only and is not sold to patients.
  • (R)-Thalidomide binds more strongly to the selective sites of MIP (molecularly imprinted polymer) compared to other forms, indicating distinct biological entities.
  • In vivo studies in F344 rats with 9L gliosarcoma tumors showed that serum and tissue concentrations of (R)-thalidomide were 40-50% greater than those of (S)-thalidomide.
  • Co-administration of BCNU or cisplatin with thalidomide did not alter the concentration enantioselectivity in rats.
Catalog No. 50-003-63556 Supplier Medchemexpress LLC Supplier No. HY14658B1MG
(R)-Thalidomide, also known as (R)-(+)-Thalidomide, is the R-enantiomer of Thalidomide, possessing psychomotor stabilizing properties. It is intended for research use only and is not sold to patients.
  • (R)-Thalidomide binds more strongly to the selective sites of MIP (molecularly imprinted polymer) compared to other forms, indicating distinct biological entities.
  • In vivo studies in F344 rats with 9L gliosarcoma tumors showed that serum and tissue concentrations of (R)-thalidomide were 40-50% greater than those of (S)-thalidomide.
  • Co-administration of BCNU or cisplatin with thalidomide did not alter the concentration enantioselectivity in rats.