Learn More
Cayman Chemical R-TrItyl glycIdyl ether 5g
SDP
Catalog No. 502950429
Encompass_Preferred
A synthetic precursor; has been used in the synthesis of glycerophospholipids, as well as compounds with antiviral and antimalarial activities
Catalog No. 50-295-0429 Supplier Cayman Chemical Supplier No. 341315 G
A synthetic precursor; has been used in the synthesis of glycerophospholipids, as well as compounds with antiviral and antimalarial activities