Learn More
Medchemexpress LLC HY-101445A 50mg Medchemexpress, (R)-Trolox CAS:53101-49-8 Purity:>98%
SDP
Catalog No. 501967848
Encompass_Preferred
Medchemexpress, HY-101445A 50mg (R)-Trolox CAS:53101-49-8 (R)-Trolox is a water soluble vitamin E analogue and a competitive tyrosinase inhibitor with a Ki value of 0.83 mM and a ID50 value of 1.88 mM. The (R)-Trolox has stronger tyrosinase affinity than the (S) enantiomer (Ki value of 0.61 mM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7848 Supplier Medchemexpress LLC Supplier No. HY101445A50MG
Add to Cart
Add to Cart
Medchemexpress, HY-101445A 50mg (R)-Trolox CAS:53101-49-8 (R)-Trolox is a water soluble vitamin E analogue and a competitive tyrosinase inhibitor with a Ki value of 0.83 mM and a ID50 value of 1.88 mM. The (R)-Trolox has stronger tyrosinase affinity than the (S) enantiomer (Ki value of 0.61 mM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.