Learn More
Medchemexpress LLC (R)-TTA-P2 | 1072018-68-8 | 100.0% | 431.37 | C21H29Cl2FN2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10035A10MM 1ML
Encompass_Preferred
(R)-TTA-P2 is the R-enantiomer of TTA-P2, a selective T-type calcium channel inhibitor used as a research reagent and experimental control. It is offered as a 10 mM solution in DMSO (1 mL) and in solid forms. Molecular formula C21H29Cl2FN2O2 and molecular weight 431.37. Enantiomeric purity is reported as 99.95% ee.
  • Supplied as 10 mM solution in DMSO, 1 mL.
  • Also available as solid material in multiple pack sizes.
  • High enantiomeric purity (99.95% ee).
  • Molecular weight 431.37 and formula C21H29Cl2FN2O2.
  • Intended for research use as an experimental control in T-type calcium channel studies.
  • Safety data sheet available for handling and storage information.
Catalog No. 50-002-13281