Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-UT-155 | 2031161-54-1 | 98.1% | 405.35 | 10 MG
SDP

Catalog No. 5000438792
Encompass_Preferred

(R)-UT-155 (compound 11) is a selective androgen receptor degrader (SARD) ligand. It is less active than its S-isomer. This compound is used in various research applications, particularly in cancer and urogenital disease studies.

  • High purity: 98.1%
  • Molecular weight: 405.35
  • Solid appearance, white to off-white color
  • Soluble in DMSO at 100 mg/mL
  • Effective antagonist activity at human androgen receptor in HEK293 cells (IC50: 0.598 μM)
  • Stable storage conditions for powder and in solvent

Catalog No. 50-004-38792 Supplier Medchemexpress LLC Supplier No. HY112895A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-UT-155 (compound 11) is a selective androgen receptor degrader (SARD) ligand. It is less active than its S-isomer. This compound is used in various research applications, particularly in cancer and urogenital disease studies.

  • High purity: 98.1%
  • Molecular weight: 405.35
  • Solid appearance, white to off-white color
  • Soluble in DMSO at 100 mg/mL
  • Effective antagonist activity at human androgen receptor in HEK293 cells (IC50: 0.598 μM)
  • Stable storage conditions for powder and in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.