Learn More
Medchemexpress LLC (R)-UT-155 | 2031161-54-1 | 98.1% | 405.35 | 100 MG
SDP
Catalog No. 5000439241
Encompass_Preferred
(R)-UT-155 (compound 11) is a selective androgen receptor degrader (SARD) ligand. It is less active than the S-isomer.
  • Selective androgen receptor degrader (SARD) ligand.
  • Less active than the S-isomer.
  • For research use only.
Catalog No. 50-004-39241 Supplier Medchemexpress LLC Supplier No. HY112895A100MG
(R)-UT-155 (compound 11) is a selective androgen receptor degrader (SARD) ligand. It is less active than the S-isomer.
  • Selective androgen receptor degrader (SARD) ligand.
  • Less active than the S-isomer.
  • For research use only.