Learn More
Medchemexpress LLC R-verapamil HCl | 38176-02-2 | MFCD00069355 | 99.4% | 491.06 g/mol | C27H39ClN2O4 | 5 MG
SDP
Catalog No. 5000223215
Encompass_Preferred
(R)-Verapamil hydrochloride is the R-enantiomer of verapamil supplied as a hydrochloride salt for research use. It acts as an L-type calcium channel blocker and a P-glycoprotein modulator, and is used in studies of calcium signaling, drug transport, and apoptosis. Supplied as a high-purity reagent suitable for in vitro and pharmacological research.
  • R enantiomer of verapamil for stereospecific studies.
  • L-type calcium channel blocker activity.
  • P-glycoprotein modulation to study drug transport.
  • High purity suitable for in vitro assays.
  • Available in small quantities for screening and mechanistic experiments.
Catalog No. 50-002-23215 Supplier Medchemexpress LLC Supplier No. HY1353365MG
(R)-Verapamil hydrochloride is the R-enantiomer of verapamil supplied as a hydrochloride salt for research use. It acts as an L-type calcium channel blocker and a P-glycoprotein modulator, and is used in studies of calcium signaling, drug transport, and apoptosis. Supplied as a high-purity reagent suitable for in vitro and pharmacological research.
  • R enantiomer of verapamil for stereospecific studies.
  • L-type calcium channel blocker activity.
  • P-glycoprotein modulation to study drug transport.
  • High purity suitable for in vitro assays.
  • Available in small quantities for screening and mechanistic experiments.