Learn More
Medchemexpress LLC (R)-verapamil hydrochloride | 38176-02-2 | MFCD00069355 | 99.4% | C27H39ClN2O4 | 10MG
SDP
Catalog No. 5000258487
Encompass_Preferred
(R)-Verapamil hydrochloride is the R-enantiomer of verapamil presented as the hydrochloride salt. It is used as a research reagent to study L-type calcium channel inhibition and modulation of drug transporters such as P-glycoprotein/MRP1. The material is supplied as a solid with high reported purity suitable for biochemical and pharmacological assays.
  • High chemical purity, 99.4% by manufacturer.
  • R-enantiomer for stereospecific pharmacological studies.
  • Solid form suitable for long-term storage and handling.
  • Suitable for studies of calcium channel activity and transporter modulation.
  • Available in small lab-scale package sizes for research use.
Catalog No. 50-002-58487 Supplier Medchemexpress LLC Supplier No. HY13533610MG
(R)-Verapamil hydrochloride is the R-enantiomer of verapamil presented as the hydrochloride salt. It is used as a research reagent to study L-type calcium channel inhibition and modulation of drug transporters such as P-glycoprotein/MRP1. The material is supplied as a solid with high reported purity suitable for biochemical and pharmacological assays.
  • High chemical purity, 99.4% by manufacturer.
  • R-enantiomer for stereospecific pharmacological studies.
  • Solid form suitable for long-term storage and handling.
  • Suitable for studies of calcium channel activity and transporter modulation.
  • Available in small lab-scale package sizes for research use.