Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-viloxazine hydrochloride | 56287-63-9 | MFCD18379655 | 99.1% | 273.76 | C13H20ClNO3 | 5MG
SDP

Catalog No. 5000236576
Encompass_Preferred

(R)-Viloxazine hydrochloride is the R-enantiomer of viloxazine hydrochloride supplied as a high-purity research chemical for laboratory pharmacology and biochemical studies. It is described as a less active enantiomer of viloxazine and is characterized by a defined molecular composition, physical form, and recommended storage conditions.

  • R-enantiomer of viloxazine hydrochloride.
  • Cas 56287-63-9.
  • Purity 99.1%.
  • Molecular formula C13H20ClNO3.
  • Molecular weight 273.76.
  • Appearance solid powder.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for human consumption.

Catalog No. 50-002-36576 Supplier Medchemexpress LLC Supplier No. HY125784A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Viloxazine hydrochloride is the R-enantiomer of viloxazine hydrochloride supplied as a high-purity research chemical for laboratory pharmacology and biochemical studies. It is described as a less active enantiomer of viloxazine and is characterized by a defined molecular composition, physical form, and recommended storage conditions.

  • R-enantiomer of viloxazine hydrochloride.
  • Cas 56287-63-9.
  • Purity 99.1%.
  • Molecular formula C13H20ClNO3.
  • Molecular weight 273.76.
  • Appearance solid powder.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.