Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-viloxazine hydrochloride | 56287-63-9 | MFCD18379655 | 99.1% | 273.76 g/mol | C13H20ClNO3 | 1MG
SDP

Catalog No. 5000239111
Encompass_Preferred

(R)-Viloxazine hydrochloride is the less active R-isomer of viloxazine hydrochloride, a norepinephrine reuptake inhibitor investigated for antidepressant activity. Supplied as a high-purity white to off-white solid for laboratory research, it is suitable for formulation and preclinical studies and is soluble in DMSO at useful concentrations.

  • High purity: 99.08% (supplier reported)
  • Molecular formula: C13H20ClNO3
  • Molecular weight: 273.76 g/mol
  • White to off-white solid form
  • Recommended DMSO solubility up to 25 mg/mL
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Available in small milligram research sizes

Catalog No. 50-002-39111 Supplier Medchemexpress LLC Supplier No. HY125784A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Viloxazine hydrochloride is the less active R-isomer of viloxazine hydrochloride, a norepinephrine reuptake inhibitor investigated for antidepressant activity. Supplied as a high-purity white to off-white solid for laboratory research, it is suitable for formulation and preclinical studies and is soluble in DMSO at useful concentrations.

  • High purity: 99.08% (supplier reported)
  • Molecular formula: C13H20ClNO3
  • Molecular weight: 273.76 g/mol
  • White to off-white solid form
  • Recommended DMSO solubility up to 25 mg/mL
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Available in small milligram research sizes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.