Learn More
Medchemexpress LLC HY-101281B 10mg Medchemexpress, (R)-VU 6008667 CAS:2097818-14-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101281B10MG
Encompass_Preferred
Medchemexpress, HY-101281B 10mg (R)-VU 6008667 CAS:2097818-14-7 (R)-VU 6008667, the less active (R)-enantiomer to VU 6008667, is devoid of M5 NAM activity (IC50>10 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7823