Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-VU 6008667 | 2097818-14-7 | 99.8% | 438.85 | C24H17ClF2N2O2 | 1 ML
SDP

Catalog No. 5000211322
Encompass_Preferred

(R)-VU 6008667 is the (R)-enantiomer of VU 6008667 supplied for research use. It is described as the less active enantiomer and lacks M5 negative allosteric modulator activity (IC50 > 10 μM). Available as a solid or as a 10 mM solution in DMSO, the product includes purity, molecular, and storage information for laboratory applications.

  • High purity (99.8%).
  • Available as solid or 10 mM solution in DMSO.
  • White to off-white solid appearance.
  • Molecular weight 438.85 and formula C24H17ClF2N2O2.
  • Storage: solid at 4°C under nitrogen; in solvent at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-11322 Supplier Medchemexpress LLC Supplier No. HY101281B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-VU 6008667 is the (R)-enantiomer of VU 6008667 supplied for research use. It is described as the less active enantiomer and lacks M5 negative allosteric modulator activity (IC50 > 10 μM). Available as a solid or as a 10 mM solution in DMSO, the product includes purity, molecular, and storage information for laboratory applications.

  • High purity (99.8%).
  • Available as solid or 10 mM solution in DMSO.
  • White to off-white solid appearance.
  • Molecular weight 438.85 and formula C24H17ClF2N2O2.
  • Storage: solid at 4°C under nitrogen; in solvent at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.