Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-VU 6008667 | 2097818-14-7 | 99.8% | 438.85 g·mol⁻¹ | C24H17ClF2N2O2 | 50 MG
SDP

Catalog No. 5000211152
Encompass_Preferred

(R)-VU 6008667 is the less-active R-enantiomer of VU 6008667, supplied as a high-purity research chemical for in vitro pharmacology and medicinal chemistry. It is reported to lack muscarinic acetylcholine receptor M5 negative allosteric modulator activity (IC50 > 10 μM) and is intended for laboratory research use only.

  • High purity: 99.8% (manufacturer listing).
  • Molecular weight: 438.85 g·mol⁻¹.
  • Chemical formula: C24H17ClF2N2O2.
  • Available in 50 mg solid packaging and solution formats.
  • Suitable as a research reagent for in vitro studies; not for human use.

Catalog No. 50-002-11152 Supplier Medchemexpress LLC Supplier No. HY101281B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-VU 6008667 is the less-active R-enantiomer of VU 6008667, supplied as a high-purity research chemical for in vitro pharmacology and medicinal chemistry. It is reported to lack muscarinic acetylcholine receptor M5 negative allosteric modulator activity (IC50 > 10 μM) and is intended for laboratory research use only.

  • High purity: 99.8% (manufacturer listing).
  • Molecular weight: 438.85 g·mol⁻¹.
  • Chemical formula: C24H17ClF2N2O2.
  • Available in 50 mg solid packaging and solution formats.
  • Suitable as a research reagent for in vitro studies; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.