Learn More
Medchemexpress LLC HY-19939S2 1mg Medchemexpress, (R)-VX-984 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19939S21MG
Encompass_Preferred
Medchemexpress, HY-19939S2 1mg (R)-VX-984 CAS: (R)-VX-984 ((R)-M9831) is the (R)-enantiomer of VX-984. VX-984 is a potent DNA-PK inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2846