Learn More
Medchemexpress LLC R-(+)-warfarin | 5543-58-8 | MFCD00272376 | 99.6% | 308.33 g·mol⁻¹ | C19H16O4 | 5 MG
SDP
Catalog No. 5000299934
Encompass_Preferred
(R)-(+)-Warfarin is the R-enantiomer of warfarin provided as a high-purity biochemical assay reagent for research use only. It is used in stereoselective metabolism and enzymatic studies, including investigations of cytochrome P450-mediated biotransformation and vitamin K-related pathways.
  • High purity suitable for analytical and assay work.
  • R-enantiomer for stereoselective metabolic studies.
  • Useful as a substrate for CYP enzymes and metabolic assays.
  • Available in small weighed quantities for research applications.
  • Molecular formula C19H16O4, molecular weight 308.33 g·mol⁻¹.
Catalog No. 50-002-99934 Supplier Medchemexpress LLC Supplier No. HYW0159985MG
(R)-(+)-Warfarin is the R-enantiomer of warfarin provided as a high-purity biochemical assay reagent for research use only. It is used in stereoselective metabolism and enzymatic studies, including investigations of cytochrome P450-mediated biotransformation and vitamin K-related pathways.
  • High purity suitable for analytical and assay work.
  • R-enantiomer for stereoselective metabolic studies.
  • Useful as a substrate for CYP enzymes and metabolic assays.
  • Available in small weighed quantities for research applications.
  • Molecular formula C19H16O4, molecular weight 308.33 g·mol⁻¹.