Learn More
Medchemexpress LLC (R)-zanubrutinib | 1691249-44-1 | MFCD31567462 | 99.0% | 471.5 g/mol | C27H29N5O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101474B25MG
Encompass_Preferred
(R)-Zanubrutinib is the R enantiomer of zanubrutinib, a selective inhibitor of Bruton's tyrosine kinase (BTK). It is provided as a high-purity research compound for biochemical assays and preclinical studies focused on BTK signaling and inhibition.
  • High purity suitable for biochemical assays.
  • Potent BTK inhibition (IC50 11 nM).
  • Available in small research pack sizes for preclinical work.
  • Useful for target validation and signaling pathway studies.
  • Provided with SDS and CAS registry information for traceability.
Catalog No. 50-002-49216