Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Zelebrudomide | 3024312-52-2 | C39H45N9O5 | 100 MG
SDP

Catalog No. 5000372503
Encompass_Preferred

(R)-Zelebrudomide ((R)-NX-2127) is an isomer of the BTK PROTAC degrader Zelebrudomide. It is composed of a BTK ligand, (S)-4-(1-(Pyrrolidin-3-ylmethyl)piperidin-4-yl)aniline, and Thalidomide 5-fluoride. This product is intended for research use only.

  • Isomer of the BTK PROTAC degrader Zelebrudomide
  • Composed of a BTK ligand, (S)-4-(1-(Pyrrolidin-3-ylmethyl)piperidin-4-yl)aniline, and Thalidomide 5-fluoride
  • For research use only
  • High purity (98.93%)

Catalog No. 50-003-72503 Supplier Medchemexpress LLC Supplier No. HY153220A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Zelebrudomide ((R)-NX-2127) is an isomer of the BTK PROTAC degrader Zelebrudomide. It is composed of a BTK ligand, (S)-4-(1-(Pyrrolidin-3-ylmethyl)piperidin-4-yl)aniline, and Thalidomide 5-fluoride. This product is intended for research use only.

  • Isomer of the BTK PROTAC degrader Zelebrudomide
  • Composed of a BTK ligand, (S)-4-(1-(Pyrrolidin-3-ylmethyl)piperidin-4-yl)aniline, and Thalidomide 5-fluoride
  • For research use only
  • High purity (98.93%)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.