Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Zelebrudomide | 3024312-52-2 | C39H45N9O5 | 5 MG
SDP

Catalog No. 5000373782
Encompass_Preferred

(R)-Zelebrudomide is an isomer of the BTK PROTAC degrader Zelebrudomide. It is also known as (R)-NX-2127 and (R)-Compound 28. This compound includes a BTK ligand 10, (S)-4-(1-(Pyrrolidin-3-ylmethyl)piperidin-4-yl)aniline, and Thalidomide 5-fluoride.

  • Purity: 98.93%
  • Molecular weight: 719.83
  • Appearance: Light yellow to yellow solid
  • Storage: 4°C, protected from light; in solvent: -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-003-73782 Supplier Medchemexpress LLC Supplier No. HY153220A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Zelebrudomide is an isomer of the BTK PROTAC degrader Zelebrudomide. It is also known as (R)-NX-2127 and (R)-Compound 28. This compound includes a BTK ligand 10, (S)-4-(1-(Pyrrolidin-3-ylmethyl)piperidin-4-yl)aniline, and Thalidomide 5-fluoride.

  • Purity: 98.93%
  • Molecular weight: 719.83
  • Appearance: Light yellow to yellow solid
  • Storage: 4°C, protected from light; in solvent: -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.