Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R-zevaquenabant | 00-00-0 | 99.2% | 548.0 g/mol | C25H21ClF3N5O2S | 5 MG
SDP

Catalog No. 5000223167
Encompass_Preferred

(R)-Zevaquenabant is the R-enantiomer of Zevaquenabant (MRI-1867), a peripherally restricted, orally bioavailable dual cannabinoid CB1 receptor and inducible nitric oxide synthase (iNOS) antagonist. Supplied as a white to off-white solid in small research quantities, it is intended for in vitro and in vivo pharmacology studies exploring CB1/iNOS pathways. Manufacturer-reported purity and enantiomeric excess accompany the product.

  • High enantiomeric purity (ee 99.58%) and chemical purity (~99.15%).
  • White to off-white solid, easy to weigh and prepare.
  • Available in small research sizes for pilot experiments.
  • Suitable for CB1 receptor and iNOS pharmacology studies.
  • Characterized by formula C25H21ClF3N5O2S and molecular weight 548.0 g/mol.

Catalog No. 50-002-23167 Supplier Medchemexpress LLC Supplier No. HY141411B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Zevaquenabant is the R-enantiomer of Zevaquenabant (MRI-1867), a peripherally restricted, orally bioavailable dual cannabinoid CB1 receptor and inducible nitric oxide synthase (iNOS) antagonist. Supplied as a white to off-white solid in small research quantities, it is intended for in vitro and in vivo pharmacology studies exploring CB1/iNOS pathways. Manufacturer-reported purity and enantiomeric excess accompany the product.

  • High enantiomeric purity (ee 99.58%) and chemical purity (~99.15%).
  • White to off-white solid, easy to weigh and prepare.
  • Available in small research sizes for pilot experiments.
  • Suitable for CB1 receptor and iNOS pharmacology studies.
  • Characterized by formula C25H21ClF3N5O2S and molecular weight 548.0 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.