Learn More
Cayman Chemical (R)-ZINC3573, Purity: ≥95%, CAS: 2089389-15-9, 50 mg
SDP
Catalog No. 502944082
Encompass_Preferred
An MRGPRX2 agonist; induces β-arrestin recruitment in a reporter assay using MRGPRX2-expressing HTLA cells and calcium mobilization in LAD 2 mast cells (EC50s = 0.74 and 1 µM, respectively); selective for MRGPRX2 over 21 other GPCRs; induces degranulation of LAD 2 mast cells
Catalog No. 50-294-4082 Supplier Cayman Chemical Supplier No. 4131250 MG
An MRGPRX2 agonist; induces β-arrestin recruitment in a reporter assay using MRGPRX2-expressing HTLA cells and calcium mobilization in LAD 2 mast cells (EC50s = 0.74 and 1 µM, respectively); selective for MRGPRX2 over 21 other GPCRs; induces degranulation of LAD 2 mast cells