Learn More
Cayman Chemical R1 Methanandamide Phosphate
SDP
Catalog No. 502890842
Encompass_Preferred
A water soluble prodrug analog of AEA; similar activity to AEA in the growth inhibition of C6 glioma cells; a structural variant of LPA
Catalog No. 50-289-0842 Supplier Cayman Chemical Supplier No. 100042811 MG
A water soluble prodrug analog of AEA; similar activity to AEA in the growth inhibition of C6 glioma cells; a structural variant of LPA