Learn More
Cayman Chemical R1 Methanandamide Phosphate
SDP
Supplier:  Cayman Chemical 1000428110 MG
Encompass_Preferred
A water soluble prodrug analog of AEA; similar activity to AEA in the growth inhibition of C6 glioma cells; a structural variant of LPA
Catalog No. 50-288-7262