Learn More
Alkali Scientific Alkali Scientific™ R1000 Cool Cube Microtube and PCR Plate Cooler
SDP
Catalog No. 503026909
Encompass_Preferred
The Alkali Scientific™ R1000 CoolCube™ Microtube and PCR Plate Cooler is designed to store samples safely on the lab bench without sample degradation due to temperature increase or fluctuation. Simply place the CoolCube in the freezer overnight and it’s ready for use, keeping samples cold, but not frozen. One side of the CoolCube will hold 36×1.5/2.0ml tubes. Flip it over, and you have a PCR workstation. This side holds one 96 well PCR plate (or 96×0.2ml tubes) and ten 1.5/2.0ml tubes.CoolCube maintains temperature at approximately 0° C for up to four hours on the bench. Maintains 0 °C for up to 4 hours Also for use as a PCR workstation Two sides, ideal for:      1.5 & 2.0ml microtubes       0.2ml PCR tubes & strips       96 well PCR plates      Size 5.6 x 5.6 x 2.5 in
Catalog No. 50-302-6909 Supplier Alkali Scientific Supplier No. RS70197
The Alkali Scientific™ R1000 CoolCube™ Microtube and PCR Plate Cooler is designed to store samples safely on the lab bench without sample degradation due to temperature increase or fluctuation. Simply place the CoolCube in the freezer overnight and it’s ready for use, keeping samples cold, but not frozen. One side of the CoolCube will hold 36×1.5/2.0ml tubes. Flip it over, and you have a PCR workstation. This side holds one 96 well PCR plate (or 96×0.2ml tubes) and ten 1.5/2.0ml tubes.CoolCube maintains temperature at approximately 0° C for up to four hours on the bench. Maintains 0 °C for up to 4 hours Also for use as a PCR workstation Two sides, ideal for:      1.5 & 2.0ml microtubes       0.2ml PCR tubes & strips       96 well PCR plates      Size 5.6 x 5.6 x 2.5 in