Learn More
Cayman Chemical R1487 (hydrochloride), Purity: ≥98%, CAS: 449808-64-4, 10 mg
SDP
Catalog No. 502991081
Encompass_Preferred
An inhibitor of p38α MAPK (IC50 = 10 nM); selective for p38α over p38β MAPK (Kds = 0.2 and 29 nM, respectively), as well as a panel of 306 additional kinases at 10 µM, but does inhibit 11 kinases by >85% at 10 µM; inhibits LPS-induced IL-1β production in isolated human whole blood (IC50 = 170 nM); inhibits LPS-induced production of TNF-α and IL-6 in rats (ED50s = 0.8 and 0.4 mg/kg); reduces yeast-induced hyperalgesia in rats (ED50 = 5.5 mg/kg)
Catalog No. 50-299-1081 Supplier Cayman Chemical Supplier No. 3147810 MG
An inhibitor of p38α MAPK (IC50 = 10 nM); selective for p38α over p38β MAPK (Kds = 0.2 and 29 nM, respectively), as well as a panel of 306 additional kinases at 10 µM, but does inhibit 11 kinases by >85% at 10 µM; inhibits LPS-induced IL-1β production in isolated human whole blood (IC50 = 170 nM); inhibits LPS-induced production of TNF-α and IL-6 in rats (ED50s = 0.8 and 0.4 mg/kg); reduces yeast-induced hyperalgesia in rats (ED50 = 5.5 mg/kg)