Learn More
CPC Scientific H-Arg-Ile-Gln-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-Gly-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HIV004A
Encompass
Peptide arised from the V3 loop of Hiv-1 IIIB gp120 efficiently blocked HIV-1 IIIB infection of several T-cell lines and normal human T cells that inhibitis syncytium formation in human cells. It has the capacity to serve as an inducer of HIV-1 specific cytotoxic T lymphocyte responses that could effectively kill virus-infected cells, thereby serving several therapeutic applications. ONE-LETTER SEQUENCE: RIQRGPGRAFVTIGKMOLECULAR FORMULA: C73H126N26O18MOLECULAR WEIGHT:1656STORAGE CONDITIONS: -20 5CRESEARCH AREA: AntimicrobialREFERENCES: R. Nehete, J. Virol., 67, 6841 (1993)
Catalog No. 50-210-9535