Learn More
Selleck Chemical LLC R16 E1973-1G
SDP
Catalog No. 5000039133
Encompass_Preferred
R16(NSC29869 Nrf2-IN-3) is an inhibitor of NRF2 (nuclear factor erythroid 2-related factor 2) that selectively binds KEAP1 mutants and restores their NRF2-inhibitory function by repairing the disrupted KEAP1/NRF2 interactions It significantly enhances the sensitivity of KEAP1-mutated tumor cells to cisplatin and gefitinib
Catalog No. 50-000-39133 Supplier Selleck Chemical LLC Supplier No. E19731G
R16(NSC29869 Nrf2-IN-3) is an inhibitor of NRF2 (nuclear factor erythroid 2-related factor 2) that selectively binds KEAP1 mutants and restores their NRF2-inhibitory function by repairing the disrupted KEAP1/NRF2 interactions It significantly enhances the sensitivity of KEAP1-mutated tumor cells to cisplatin and gefitinib