Learn More
Cayman Chemical R18 (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502909541
Encompass_Preferred
A peptide inhibitor of the 14-3-3-Raf-1 protein-protein interaction; binds to 14-3-3τ, -β, and -ζ (Kds = 7.1, 7, and 8.9 nM, respectively) and inhibits 14-3-3τ binding to Raf-1 in a filter binding assay at 5 nM; inhibits 14-3-3ζ-mediated protection of Raf-1 from phosphatase-induced inactivation
Catalog No. 50-290-9541 Supplier Cayman Chemical Supplier No. 366955 MG
A peptide inhibitor of the 14-3-3-Raf-1 protein-protein interaction; binds to 14-3-3τ, -β, and -ζ (Kds = 7.1, 7, and 8.9 nM, respectively) and inhibits 14-3-3τ binding to Raf-1 in a filter binding assay at 5 nM; inhibits 14-3-3ζ-mediated protection of Raf-1 from phosphatase-induced inactivation