Learn More
Cayman Chemical (R)-2-(3-(2-Ethoxyphenoxy)piperidin-1-yl)pyrimidine-5-carboxylic Acid, Purity: ≥95%, CAS: 1809064-89-8, 10 mg
SDP
Catalog No. 502918515
Encompass_Preferred
An intermediate in the synthesis of ervogastat
Catalog No. 50-291-8515 Supplier Cayman Chemical Supplier No. 3925710 MG
An intermediate in the synthesis of ervogastat