Learn More
Cole-Parmer IKA 3372000 Support Clamp (r 200) For T-10 Basic Homogenizer
Catalog No. NC1343901 Shop All Cole-Parmer Products
Encompass
Diameter of extension arm: 8 mm, Length of extension arm: 130 mm, R 200 Clamp for fastening the T 10 basic to the stand R 104.
Catalog No. NC1343901 Supplier Cole-Parmer Supplier No. 0472080
Diameter of extension arm: 8 mm, Length of extension arm: 130 mm, R 200 Clamp for fastening the T 10 basic to the stand R 104.