Learn More
Medchemexpress LLC HY-122219 5mg , R243 CAS:688352-84-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1222195MG
Encompass_Preferred
HY-122219 5mg R243 CAS: 688352-84-3 R243 is a potent and selective CCR8 antagonist. R243 inhibits CCL1/CCR8 interaction and inhibits CCR8 signaling and chemotaxis. R243 has antinociceptive and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1322