Learn More
Polysciences, Inc. Coomassie Blue R250, C.I. 42660, 6104-59-2, MFCD00041762, 100 G
SDP
Catalog No. NC9867877
Encompass_Preferred
BRILLIANT BLUE R, Molecular Formula: C45 H44 N3 O7 S2 . Na, Molecular Weight: 825.979, Synonyms: Coomassie blue R; Brilliant blue R250; Acid blue 83; C.I. 42660. Coomassie Blue dyes are commonly used stains for the detection of proteins bands following gel electrophoresis. The use of Coomassie R-250 is the more common as it can detect as little as 0.1ug of protein.
Catalog No. NC9867877 Supplier Polysciences, Inc. Supplier No. 00352100
BRILLIANT BLUE R, Molecular Formula: C45 H44 N3 O7 S2 . Na, Molecular Weight: 825.979, Synonyms: Coomassie blue R; Brilliant blue R250; Acid blue 83; C.I. 42660. Coomassie Blue dyes are commonly used stains for the detection of proteins bands following gel electrophoresis. The use of Coomassie R-250 is the more common as it can detect as little as 0.1ug of protein.