Learn More
Alkali Scientific R2A Agar 10mL 60x115mm Dish, 10/Pk
SDP
Catalog No. 5000663321
Encompass_Preferred
R2A Agar, 10mL, 60x115mm Dish, 10/Pk is a low nutrient culture medium formulated for the cultivation and enumeration of heterotrophic bacteria in treated water systems, including potable, bottled, and pharmaceutical grade water. Its reduced nutrient composition encourages the recovery of slow growing or stressed microorganisms that may be inhibited on richer media. The 10mL fill in a compact 60x115mm dish supports consistent sample distribution and efficient incubation in environmental monitoring workflows. Commonly used for heterotrophic plate counts in water testing, the sterile ready to use plates support long incubation periods and reliable microbial recovery in cleanroom validation, pharmaceutical water system monitoring, and regulated laboratory environments.
Catalog No. 50-006-63321 Supplier Alkali Scientific Supplier No. R2AP01
R2A Agar, 10mL, 60x115mm Dish, 10/Pk is a low nutrient culture medium formulated for the cultivation and enumeration of heterotrophic bacteria in treated water systems, including potable, bottled, and pharmaceutical grade water. Its reduced nutrient composition encourages the recovery of slow growing or stressed microorganisms that may be inhibited on richer media. The 10mL fill in a compact 60x115mm dish supports consistent sample distribution and efficient incubation in environmental monitoring workflows. Commonly used for heterotrophic plate counts in water testing, the sterile ready to use plates support long incubation periods and reliable microbial recovery in cleanroom validation, pharmaceutical water system monitoring, and regulated laboratory environments.