Learn More
Bioworld R2A Agar, 1 kg
Catalog No. 50254358
Encompass_Preferred
R2A AgarLow nutrient medium for plate detection of heterotropic bacteria in water Low concentrations of carbon and energy sources enhance the recovery of stressed bacteriaWorking Concentration: 18.2 g/LPrepared Appearance: light amber agarSoluble in boiling waterContains: Casein Digest Peptone Peptic Digest of Animal Tissue Magnesium Sulfate Heptahydrate Sodium Pyruvate Casein Acid Hydrolysafe Dipotassium Phosphate Yeast Extract Soluble Starch Agar Glucose
Catalog No. 50-254-358 Supplier Bioworld Supplier No. 306200682
R2A AgarLow nutrient medium for plate detection of heterotropic bacteria in water Low concentrations of carbon and energy sources enhance the recovery of stressed bacteriaWorking Concentration: 18.2 g/LPrepared Appearance: light amber agarSoluble in boiling waterContains: Casein Digest Peptone Peptic Digest of Animal Tissue Magnesium Sulfate Heptahydrate Sodium Pyruvate Casein Acid Hydrolysafe Dipotassium Phosphate Yeast Extract Soluble Starch Agar Glucose