Learn More
MilliporeSigma R2A Agar, 120/pk
Catalog No. NC1276509
Encompass
R2A Agar plates are designed for the determination of the total microbial count in water for injections in bulk, highly purified water, and purified water in bulk.
Catalog No. NC1276509 Supplier MilliporeSigma Supplier No. 1460750120
R2A Agar plates are designed for the determination of the total microbial count in water for injections in bulk, highly purified water, and purified water in bulk.