Learn More
Alkali Scientific R2A Agar, 500 G
SDP
Catalog No. 5000661721
Encompass_Preferred
R2A Agar (Low Nutrient Media) is designed for the growth and recovery of heterotrophic bacteria in water quality testing and environmental microbiology. The 500g package provides a sufficient volume for medium-scale testing, allowing for the detection and enumeration of slow-growing bacteria that may not thrive on standard rich media. This agar is particularly beneficial in testing treated potable water and pharmaceutical process water. The low-nutrient formulation encourages the recovery of stressed bacteria without promoting rapid overgrowth, making it ideal for heterotrophic plate count (HPC) procedures and microbial analysis of water and other environmental samples.
Catalog No. 50-006-61721 Supplier Alkali Scientific Supplier No. RAA002CP
R2A Agar (Low Nutrient Media) is designed for the growth and recovery of heterotrophic bacteria in water quality testing and environmental microbiology. The 500g package provides a sufficient volume for medium-scale testing, allowing for the detection and enumeration of slow-growing bacteria that may not thrive on standard rich media. This agar is particularly beneficial in testing treated potable water and pharmaceutical process water. The low-nutrient formulation encourages the recovery of stressed bacteria without promoting rapid overgrowth, making it ideal for heterotrophic plate count (HPC) procedures and microbial analysis of water and other environmental samples.