Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp R2A Agar, 500 Grams
SDP

Catalog No. 50488984
Encompass_Preferred

Low nutrient medium for plate detection of heterotropic bacteria in water. Low concentrations of carbon and energy sources enhance the recovery of stressed bacteria.

Formula per liter

Casein Digest Peptone: 0.25g
Peptic Digest of Animal Tissue: 0.25g
Magnesium Sulfate Heptahydrate: 0.05g
Sodium Pyruvate: 0.3g
Casein Acid Hydrolysate: 0.5g
Dipotassium Phosphate: 0.3g
Yeast Extract: 0.5g
Soluble Starch: 0.5g
Agar: 15.0g
Glucose: 0.5g

Catalog No. 50-488-984 Supplier Research Products International Corp Supplier No. R24200500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Low nutrient medium for plate detection of heterotropic bacteria in water. Low concentrations of carbon and energy sources enhance the recovery of stressed bacteria.

Formula per liter

Casein Digest Peptone: 0.25g
Peptic Digest of Animal Tissue: 0.25g
Magnesium Sulfate Heptahydrate: 0.05g
Sodium Pyruvate: 0.3g
Casein Acid Hydrolysate: 0.5g
Dipotassium Phosphate: 0.3g
Yeast Extract: 0.5g
Soluble Starch: 0.5g
Agar: 15.0g
Glucose: 0.5g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.