Learn More
Hach Company R2A Agar, Dehydrated 500 g
Supplier:  Hach Company 2281034
Encompass
Low nutrient medium for enumeration and cultivation of bacteria from potable water. Used for pour or spread plate and MF methods; described in the 16th edition of Standard Methods. Recommended for predicting nitrification more accurately in potable systems using chloramines as a secondary disinfectant.
Catalog No. 50-199-5720