Learn More
MOLTOX Molecular Toxicology Inc R2A Agar, 20 mL/plate, 20 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948627
Encompass_Preferred
R2A Agar is recommended for use in the enumeration of heterotrophic bacteria in drinking or tap water. This type of bacteria is slow growing and would normally be overrun by the more fast growing bacteria. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-627 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S13
R2A Agar is recommended for use in the enumeration of heterotrophic bacteria in drinking or tap water. This type of bacteria is slow growing and would normally be overrun by the more fast growing bacteria. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.