Learn More
Teknova R2A Agar Plates.100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388023
Encompass
0.025% Proteose Peptone #3,0.025% Tryptone,0.05% Casamino Acids,0.05% Yeast Extract,0.03% Sodium Pyruvate,0.05% Soluble Starch,0.03% Potassium Phosphate Dibasic,0.005% Magnesium Sulfate Heptahydrate,2.00% Nobel Agar
Catalog No. 50-238-8023 Supplier Teknova Supplier No. R1035
0.025% Proteose Peptone #3,0.025% Tryptone,0.05% Casamino Acids,0.05% Yeast Extract,0.03% Sodium Pyruvate,0.05% Soluble Starch,0.03% Potassium Phosphate Dibasic,0.005% Magnesium Sulfate Heptahydrate,2.00% Nobel Agar