Learn More
NEOGEN R2A Broth, 500 g
SDP
Supplier:  NEOGEN 700004593
Encompass_Preferred
R2A Broth is for use with membrane methods for the enumeration of bacteria from water samples, and is not intended for use in the diagnosis of disease or other conditions in humans. A medium of low nutritional content, for use with membrane methods for the enumeration of bacteria from water samples.For more information visit https://www.neogen.com/solutions/Microbiology/r2a-broth
Catalog No. 50-201-5684