Learn More
Teknova R2A Broth Mix. 500g.
SDP
Catalog No. 502388058
Encompass
0.050% Proteose Peptone #3, 0.050% Casamino Acids,0.050% Yeast Extract, 0.050% Glucose, 0.050% Starch, 0.030% Sodium Pyruvate, 0.030% Dipotassium Phosphate, 0.005% Magnesium Sulfate
Catalog No. 50-238-8058 Supplier Teknova Supplier No. R0005
0.050% Proteose Peptone #3, 0.050% Casamino Acids,0.050% Yeast Extract, 0.050% Glucose, 0.050% Starch, 0.030% Sodium Pyruvate, 0.030% Dipotassium Phosphate, 0.005% Magnesium Sulfate