Learn More
Bioworld R2B Broth, 500 g
Catalog No. 50254359
Encompass_Preferred
R2B BrothLow nutrient medium for plate detection of heterotropic bacteria in water Low concentrations of carbon and energy sources enhance the recovery of stressed bacteriaIdeal for membrane filtration methodspH: 7.2Working Concentration: 3.6 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber brothSoluble in waterContains (g/L): Casein Digest Peptone : 0.25g Peptic Digest of Animal Tissue: 0..25g Magnesium Sulfate Heptahydrate: 0.5g Sodium Pyruvate: 0.3g Casein Acid Hydrolysafe: 0.5g Dipotassium Phosphate: 0.3 Yeast Extract: 0.5g Soluble Starch: 0.5g Glucose: 0.5g
Catalog No. 50-254-359 Supplier Bioworld Supplier No. 306200691
R2B BrothLow nutrient medium for plate detection of heterotropic bacteria in water Low concentrations of carbon and energy sources enhance the recovery of stressed bacteriaIdeal for membrane filtration methodspH: 7.2Working Concentration: 3.6 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber brothSoluble in waterContains (g/L): Casein Digest Peptone : 0.25g Peptic Digest of Animal Tissue: 0..25g Magnesium Sulfate Heptahydrate: 0.5g Sodium Pyruvate: 0.3g Casein Acid Hydrolysafe: 0.5g Dipotassium Phosphate: 0.3 Yeast Extract: 0.5g Soluble Starch: 0.5g Glucose: 0.5g