Learn More
Alkali Scientific R2B Broth, 500 Grams
SDP
Supplier:  Alkali Scientific RBB001CP
Encompass_Preferred
R2B Broth (Low Nutrient Media) is a microbiological growth medium formulated to support the recovery and enumeration of slow-growing or oligotrophic bacteria from water and environmental samples. This 500g package provides a concentrated solution for preparing the broth, which is particularly useful in water quality testing, environmental microbiology, and heterotrophic plate count (HPC) procedures. The low-nutrient formulation helps to prevent rapid bacterial overgrowth, enabling the recovery of stressed organisms that would otherwise be missed using nutrient-rich media. It is often used in combination with R2A Agar to ensure complete recovery of microbial populations from water and pharmaceutical process systems.
Catalog No. 50-006-61912