Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich R2r Monolayer Large Grain Cvd Graphene On Silicon Wafer 6 In Diameter

Catalog No. 9201341EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-013-41EA 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 92-013-41EA Supplier Sigma Aldrich Supplier No. 9201341EA
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Roll-to-roll, high-quality, monolayer CVD graphene with large grain size (∼110mum^2) on 6 inch silicon wafer.Our Roll-to-Roll CVD graphene products are true monolayer high quality graphene, fabricated inside a Cleanroom, heavily monitored and QC to assure high reproducibility.The roll-to-roll process allows continuous, large scale graphene production. This large grain size graphene product on silicon wafer is ready to use, helps you minimize process time, and increase success rate. This product with low sheet resistance would enable unmatched reproducibility and allow high performance for CVD graphene based FET, CVD graphene based sensors, and heterostructure based micro/nano electronics.Application examples:Ultrafast TransistorOptical devicesBio/Gas SensorTransparent ElectrodeFlexible DisplaySmart CoatingThermal managementProduct of LG Electronics,

Specifications

CAS 4343-9-8--0
Molecular Formula C
Linear Formula C
Quantity 1 Ea.
Synonym Roll-to-Roll graphene
Molecular Weight (g/mol) 12.01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.