Learn More
Cole-Parmer Argos Technologies PolarSafe Cardboard Freezer Box, 5-1/4" x 5-1/4" x 3"; without Divider
Catalog No. NC1480443 Shop All Cole-Parmer Products
Encompass_Preferred
Create customized storage box configurations with these freezer boxes. A snug-fitting lid prevents accidental separation if dropped. The boxes can be used in mechanical freezers or liquid nitrogen tanks up to 196C. They are crafted of sturdy cardboard with moisture-repellent coating and designed with small holes on the bottom to allow for drainage. A divider to customize storage needs can be purchased separately.
Catalog No. NC1480443 Supplier Cole-Parmer Supplier No. 0675577
Create customized storage box configurations with these freezer boxes. A snug-fitting lid prevents accidental separation if dropped. The boxes can be used in mechanical freezers or liquid nitrogen tanks up to 196C. They are crafted of sturdy cardboard with moisture-repellent coating and designed with small holes on the bottom to allow for drainage. A divider to customize storage needs can be purchased separately.