Learn More
Cole-Parmer Argos Technologies PolarSafe Cardboard Freezer Box, 5-1/4" x 5-1/4" x 2", with 100-Place Divider
Supplier:  Cole-Parmer 0675573/DUP
Encompass_Preferred
Create customized storage box configurations with these freezer boxes. A snug-fitting lid prevents accidental separation if dropped. The boxes can be used in mechanical freezers or liquid nitrogen tanks up to 196C. They are crafted of sturdy cardboard with moisture-repellent coating and designed with small holes on the bottom to allow for drainage. Samples are kept organized using the boxs numerical grid.
Catalog No. NC1480447