Learn More
MilliporeSigma R3A Agar - LI 30ml
Catalog No. NC1677496
Encompass
Application: Subculture microorganisms, Primary isolation of aquatic microorganisms, Synonym: Reasoner's 3A agar, Packaging: 90 mm settle plate with 30 ml filling volume (20 plates per box) for long incubation (LI)
Catalog No. NC1677496 Supplier MilliporeSigma Supplier No. 1460740020
Application: Subculture microorganisms, Primary isolation of aquatic microorganisms, Synonym: Reasoner's 3A agar, Packaging: 90 mm settle plate with 30 ml filling volume (20 plates per box) for long incubation (LI)