Learn More
Aviva Systems Biology Corp R3HDM2 Antibody - middle region (ARP55106_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55106_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against R3HDM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human R3HDM2. Alternate names: KIAA1002, PR01365ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61425