Learn More
Cayman Chemical R3hydroxy MyrIstIc AcId 10mg
SDP
Catalog No. 502967520
Encompass_Preferred
A form of the myristic acid that is found in the lipid A component of some Gram negative bacteria, including Escherichia, Haemophilus, Actinobacillus, and Campylobacter species.
Catalog No. 50-296-7520 Supplier Cayman Chemical Supplier No. 1687110 MG
A form of the myristic acid that is found in the lipid A component of some Gram negative bacteria, including Escherichia, Haemophilus, Actinobacillus, and Campylobacter species.