Learn More
Cayman Chemical (R)-3-hydroxy Myristic Acid, Purity: ≥98%, CAS: 28715-21-1, 25 mg
SDP
Catalog No. 502966255
Encompass_Preferred
A form of the myristic acid that is found in the lipid A component of some Gram negative bacteria, including Escherichia, Haemophilus, Actinobacillus, and Campylobacter species.
Catalog No. 50-296-6255 Supplier Cayman Chemical Supplier No. 1687125 MG
A form of the myristic acid that is found in the lipid A component of some Gram negative bacteria, including Escherichia, Haemophilus, Actinobacillus, and Campylobacter species.